Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605660 MIM: 602306 MIM: 601962 MIM: 611440 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6834 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR6834 - microRNA 6834 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6873 - microRNA 6873 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PFDN6 - prefoldin subunit 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RGL2 - ral guanine nucleotide dissociation stimulator like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243738.1 | 2156 | Missense Mutation | CCC,CTC | P,L 516 | NP_001230667.1 | |
NM_004761.4 | 2156 | Missense Mutation | CCC,CTC | P,L 598 | NP_004752.1 | |
XM_011514781.1 | 2156 | Missense Mutation | CCC,CTC | P,L 491 | XP_011513083.1 | |
XM_011514785.2 | 2156 | Intron | XP_011513087.1 |
TAPBP - TAP binding protein (tapasin) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR46 - WD repeat domain 46 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |