Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACGAGGTATGACCCTGCTTGTGGC[C/T]GCTCATTCCCGAGGATATCAAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300314 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAF7L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TAF7L - TATA-box binding protein associated factor 7 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168474.1 | 780 | Missense Mutation | AGC,GGC | S,G 222 | NP_001161946.1 | |
NM_024885.3 | 780 | Missense Mutation | AGC,GGC | S,G 308 | NP_079161.3 | |
XM_005262152.1 | 780 | Missense Mutation | AGC,GGC | S,G 222 | XP_005262209.1 | |
XM_006724664.1 | 780 | Missense Mutation | AGC,GGC | S,G 308 | XP_006724727.1 |
Set Membership: |
HapMap |