Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGCTGGGTGATGATCAGAGTAC[A/G]CCCAAGACCGGCCTCCTGATAATCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300892 MIM: 300341 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00850 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LINC00850 - long intergenic non-protein coding RNA 850 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAGEA8 - MAGE family member A8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166400.1 | 1118 | Silent Mutation | ACA,ACG | T,T 199 | NP_001159872.1 | |
NM_001166401.1 | 1118 | Silent Mutation | ACA,ACG | T,T 199 | NP_001159873.1 | |
NM_005364.4 | 1118 | Silent Mutation | ACA,ACG | T,T 199 | NP_005355.2 | |
XM_017029520.1 | 1118 | Silent Mutation | ACA,ACG | T,T 199 | XP_016885009.1 |
MAGEA8-AS1 - MAGEA8 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |