Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATGCACAAATCCTTCCCTACTAT[C/T]CCTACAACCACAGCATGGGGCAGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605771 MIM: 607442 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX7A2L PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
COX7A2L - cytochrome c oxidase subunit 7A2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319036.1 | Intron | NP_001305965.1 | ||||
NM_001319037.1 | Intron | NP_001305966.1 | ||||
NM_001319038.1 | Intron | NP_001305967.1 | ||||
NM_001319040.1 | Intron | NP_001305969.1 | ||||
NM_004718.3 | Intron | NP_004709.2 |
EML4 - echinoderm microtubule associated protein like 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |