Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAATTCCCAACATGGACAAACCA[C/T]GCAAAGAAAATGAAGAAGAGCCGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300237 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCEAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCEAL1 - transcription elongation factor A like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006639.1 | 175 | Missense Mutation | CGC,TGC | R,C 5 | NP_001006640.1 | |
NM_001006640.1 | 175 | Missense Mutation | CGC,TGC | R,C 5 | NP_001006641.1 | |
NM_004780.2 | 175 | Missense Mutation | CGC,TGC | R,C 5 | NP_004771.2 |