Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCGTACTGCCCGCCGCCCGAGCC[C/G]TGCGAGGGCCGATAGCGAGGGTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604014 MIM: 600923 MIM: 610554 MIM: 604729 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B4GALT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B4GALT3 - beta-1,4-galactosyltransferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UFC1 - ubiquitin-fold modifier conjugating enzyme 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP21 - ubiquitin specific peptidase 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014443.2 | 74 | Intron | NP_001014443.1 | |||
NM_001319848.1 | 74 | Intron | NP_001306777.1 | |||
NM_012475.4 | 74 | Intron | NP_036607.3 | |||
XM_011509414.1 | 74 | Intron | XP_011507716.1 | |||
XM_017001036.1 | 74 | Intron | XP_016856525.1 | |||
XM_017001037.1 | 74 | Intron | XP_016856526.1 | |||
XM_017001038.1 | 74 | Intron | XP_016856527.1 | |||
XM_017001039.1 | 74 | Intron | XP_016856528.1 | |||
XM_017001040.1 | 74 | Intron | XP_016856529.1 | |||
XM_017001041.1 | 74 | Intron | XP_016856530.1 | |||
XM_017001042.1 | 74 | Intron | XP_016856531.1 |