Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAAATGAGTGGAAATGACCCCAG[C/T]GCCTGGAGCTGGGGAGGTATGTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604014 MIM: 600923 MIM: 610554 MIM: 604729 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B4GALT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B4GALT3 - beta-1,4-galactosyltransferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UFC1 - ubiquitin-fold modifier conjugating enzyme 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP21 - ubiquitin specific peptidase 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |