Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTATCCTTTAGAAGAATGTTATG[C/G]TGTCTTCATATCTAATGAGAGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDYHV1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDYHV1 - WDYHV motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283024.1 | 204 | UTR 5 | NP_001269953.1 | |||
NM_001283027.1 | 204 | UTR 5 | NP_001269956.1 | |||
NM_018024.2 | 204 | Missense Mutation | GCT,GGT | A,G 52 | NP_060494.1 | |
XM_006716597.3 | 204 | Missense Mutation | GCT,GGT | A,G 52 | XP_006716660.1 | |
XM_011517147.1 | 204 | Missense Mutation | GCT,GGT | A,G 52 | XP_011515449.1 | |
XM_011517151.2 | 204 | UTR 5 | XP_011515453.1 | |||
XM_017013601.1 | 204 | Missense Mutation | GCT,GGT | A,G 52 | XP_016869090.1 | |
XM_017013602.1 | 204 | UTR 5 | XP_016869091.1 | |||
XM_017013603.1 | 204 | UTR 5 | XP_016869092.1 |