Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGAGGTCTCTGAGTGATGAACT[C/T]GAAGCAGCAGGGCCGTGGGGCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611625 MIM: 612107 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GID8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GID8 - GID complex subunit 8 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC17A9 - solute carrier family 17 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302643.1 | Intron | NP_001289572.1 | ||||
NM_022082.3 | Intron | NP_071365.3 | ||||
XM_011528978.2 | Intron | XP_011527280.1 |