Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATGGTGCCTACAACACGTCAGC[C/T]TTTGGAATTGAGCTGCTTCCTCGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603046 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF139 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF139 - ring finger protein 139 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007218.3 | 682 | Silent Mutation | GCC,GCT | A,A 113 | NP_009149.2 | |
XM_017013002.1 | 682 | UTR 5 | XP_016868491.1 |
TATDN1 - TatD DNase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |