Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGTCAGAAGCGGCTTCCCTACGT[C/T]CCAGAGCCCTATTACCCGGAATCTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615618 MIM: 615534 | |||||||||||||||||||||||
Literature Links: |
POGLUT1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
POGLUT1 - protein O-glucosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIMMDC1 - translocase of inner mitochondrial membrane domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016589.3 | 344 | Silent Mutation | GTC,GTT | V,V 45 | NP_057673.2 | |
XM_017006556.1 | 344 | Silent Mutation | GTC,GTT | V,V 45 | XP_016862045.1 |