Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCCACATCTGACCCACCTCGCC[A/G]GTCTCAGGTGCTGCGTTGCCAGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191350 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2CD2L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2CD2L - C2CD2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290474.1 | Intron | NP_001277403.1 | ||||
NM_014807.4 | Intron | NP_055622.3 | ||||
XM_006718948.3 | Intron | XP_006719011.1 | ||||
XM_011543076.2 | Intron | XP_011541378.1 | ||||
XM_017018625.1 | Intron | XP_016874114.1 | ||||
XM_017018626.1 | Intron | XP_016874115.1 | ||||
XM_017018627.1 | Intron | XP_016874116.1 | ||||
XM_017018628.1 | Intron | XP_016874117.1 |
DPAGT1 - dolichyl-phosphate N-acetylglucosaminephosphotransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |