Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATCCCGCATCCGGCATCCCAGC[A/G]GCCGGGCATGTAGCAGCGGCAGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107986255 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107986255 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017008890.1 | 19 | UTR 5 | XP_016864379.1 |
STK32B - serine/threonine kinase 32B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306082.1 | 19 | UTR 5 | NP_001293011.1 | |||
NM_018401.2 | 19 | UTR 5 | NP_060871.1 | |||
XM_005247982.3 | 19 | Intron | XP_005248039.1 | |||
XM_005247983.4 | 19 | Intron | XP_005248040.1 | |||
XM_011513497.1 | 19 | UTR 5 | XP_011511799.1 | |||
XM_011513501.2 | 19 | Intron | XP_011511803.1 | |||
XM_011513504.1 | 19 | Intron | XP_011511806.1 | |||
XM_017008374.1 | 19 | Intron | XP_016863863.1 | |||
XM_017008375.1 | 19 | Intron | XP_016863864.1 | |||
XM_017008376.1 | 19 | Intron | XP_016863865.1 | |||
XM_017008377.1 | 19 | Intron | XP_016863866.1 |