Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTTGGAGTAATCCACAGAATAC[A/G]CTCCTTCTTCCTCAGTCACAATGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600935 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNJ8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNJ8 - potassium voltage-gated channel subfamily J member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004982.3 | 1316 | Missense Mutation | GCG,GTG | A,V 334 | NP_004973.1 | |
XM_005253358.4 | 1316 | Missense Mutation | GCG,GTG | A,V 334 | XP_005253415.1 | |
XM_017019283.1 | 1316 | Missense Mutation | GCG,GTG | A,V 334 | XP_016874772.1 | |
XM_017019284.1 | 1316 | Missense Mutation | GCG,GTG | A,V 334 | XP_016874773.1 |