Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAAGGAAGGGACTACCAACCACC[C/T]GCTGCTAGCCACAGGGCTGGGGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165161 MIM: 600538 MIM: 606034 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JUNB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JUNB - JunB proto-oncogene, AP-1 transcription factor subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRDX2 - peroxiredoxin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005809.5 | Intron | NP_005800.3 |
RNASEH2A - ribonuclease H2 subunit A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |