Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCCCAGGCCCCTGTACCTCACAC[T/C]TGTTCTCCTGCCCCTCTGCATACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609912 MIM: 610560 MIM: 600823 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KAT8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KAT8 - lysine acetyltransferase 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRSS36 - protease, serine 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258290.1 | 2326 | Missense Mutation | AAG,AGG | K,R 756 | NP_001245219.1 | |
NM_001258291.1 | 2326 | Intron | NP_001245220.1 | |||
NM_173502.4 | 2326 | Missense Mutation | AAG,AGG | K,R 761 | NP_775773.2 | |
XM_011545758.2 | 2326 | Missense Mutation | AAG,AGG | K,R 536 | XP_011544060.1 | |
XM_017022968.1 | 2326 | Missense Mutation | AAA,AGA | K,R 761 | XP_016878457.1 | |
XM_017022969.1 | 2326 | Intron | XP_016878458.1 | |||
XM_017022970.1 | 2326 | Intron | XP_016878459.1 | |||
XM_017022971.1 | 2326 | Intron | XP_016878460.1 |
PRSS8 - protease, serine 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |