Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGTGGATTGCTCAGTCCCTATC[A/G]TATTAGAGCCACTGAGGATTGAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612686 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC283278 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC283278 - pleckstrin homology domain-containing family A member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017018657.1 | Intron | XP_016874146.1 | ||||
XM_017018658.1 | Intron | XP_016874147.1 |
PLEKHA7 - pleckstrin homology domain containing A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175058.4 | Intron | NP_778228.3 | ||||
XM_017017240.1 | Intron | XP_016872729.1 | ||||
XM_017017241.1 | Intron | XP_016872730.1 | ||||
XM_017017242.1 | Intron | XP_016872731.1 | ||||
XM_017017243.1 | Intron | XP_016872732.1 |