Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCCGGGCTCTTGGTACCTCAGC[G/T]CGAGCGCCAGGCGTCCGGCCGCCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603465 MIM: 601754 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC45 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDC45 - cell division cycle 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178010.2 | 114 | UTR 5 | NP_001171481.1 | |||
NM_001178011.2 | 114 | UTR 5 | NP_001171482.1 | |||
NM_003504.4 | 114 | UTR 5 | NP_003495.1 | |||
XM_005261285.2 | 114 | UTR 5 | XP_005261342.1 | |||
XM_005261286.3 | 114 | UTR 5 | XP_005261343.1 | |||
XM_011530415.1 | 114 | UTR 5 | XP_011528717.1 | |||
XM_011530416.1 | 114 | Intron | XP_011528718.1 | |||
XM_011530417.2 | 114 | UTR 5 | XP_011528719.1 | |||
XM_011530418.2 | 114 | UTR 5 | XP_011528720.1 | |||
XM_017028966.1 | 114 | Intron | XP_016884455.1 | |||
XM_017028967.1 | 114 | Intron | XP_016884456.1 |
UFD1L - ubiquitin fusion degradation 1 like (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035247.2 | 114 | Intron | NP_001030324.2 | |||
NM_005659.6 | 114 | Intron | NP_005650.2 | |||
XM_011530361.2 | 114 | Intron | XP_011528663.1 |
Set Membership: |
HapMap |