Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCTTCTGCTTCTTTGTCTTCGG[C/T]GCCGTCTTGTTGTGTGTGGGAGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105379030 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105379030 - uncharacterized LOC105379030 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM171 - transmembrane protein 171 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161342.2 | 291 | Silent Mutation | GGC,GGT | G,G 29 | NP_001154814.1 | |
NM_173490.7 | 291 | Silent Mutation | GGC,GGT | G,G 29 | NP_775761.4 | |
XM_011543156.1 | 291 | Silent Mutation | GGC,GGT | G,G 29 | XP_011541458.1 |