Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCACTCACGGAGCCCACCAGGGA[A/G]ATGACCAGGTCCAGGCGGGGGATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608331 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC36A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC36A2 - solute carrier family 36 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181776.2 | 1259 | Silent Mutation | ATC,ATT | I,I 407 | NP_861441.2 | |
XM_005268377.3 | 1259 | Intron | XP_005268434.1 | |||
XM_006714756.3 | 1259 | Silent Mutation | ATC,ATT | I,I 374 | XP_006714819.1 | |
XM_017009083.1 | 1259 | Intron | XP_016864572.1 | |||
XM_017009084.1 | 1259 | Silent Mutation | ATC,ATT | I,I 209 | XP_016864573.1 |