Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCGTGATTTAAAGAGCTAAGTAT[C/T]TCTACCGTATTTTTCAAGCCAGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611704 | ||||||||||||||||||||
Literature Links: |
TMPRSS11A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMPRSS11A - transmembrane protease, serine 11A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114387.1 | 1609 | UTR 3 | NP_001107859.1 | |||
NM_182606.3 | 1609 | UTR 3 | NP_872412.3 |