Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCACAAGCATGCTCTCAAGCCCT[C/T]GCTCCACAAGAGCTCTGATCGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608256 MIM: 606565 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCN4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCN4B - sodium voltage-gated channel beta subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142348.1 | 3479 | UTR 3 | NP_001135820.1 | |||
NM_001142349.1 | 3479 | UTR 3 | NP_001135821.1 | |||
NM_174934.3 | 3479 | UTR 3 | NP_777594.1 |
TMPRSS4 - transmembrane protease, serine 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |