Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAACGCTCCCTAGCAAATAAGGA[G/T]AGAATTAGAAAGATTCAAGAGAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
47 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611609 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIPA1L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SIPA1L2 - signal induced proliferation associated 1 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020808.4 | Intron | NP_065859.3 | ||||
XM_005273211.2 | Intron | XP_005273268.1 | ||||
XM_005273212.4 | Intron | XP_005273269.1 | ||||
XM_005273213.4 | Intron | XP_005273270.1 | ||||
XM_011544243.2 | Intron | XP_011542545.1 | ||||
XM_017001896.1 | Intron | XP_016857385.1 | ||||
XM_017001897.1 | Intron | XP_016857386.1 | ||||
XM_017001898.1 | Intron | XP_016857387.1 |
Set Membership: |
HapMap |