Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300374 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH3KBP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SH3KBP1 - SH3 domain containing kinase binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024666.2 | Intron | NP_001019837.1 | ||||
NM_001184960.1 | Intron | NP_001171889.1 | ||||
NM_031892.2 | Intron | NP_114098.1 | ||||
XM_005274494.2 | Intron | XP_005274551.1 | ||||
XM_011545498.2 | Intron | XP_011543800.1 | ||||
XM_011545499.2 | Intron | XP_011543801.1 | ||||
XM_011545500.2 | Intron | XP_011543802.1 | ||||
XM_011545502.2 | Intron | XP_011543804.1 | ||||
XM_011545503.2 | Intron | XP_011543805.1 | ||||
XM_017029459.1 | Intron | XP_016884948.1 | ||||
XM_017029460.1 | Intron | XP_016884949.1 | ||||
XM_017029461.1 | Intron | XP_016884950.1 | ||||
XM_017029462.1 | Intron | XP_016884951.1 | ||||
XM_017029463.1 | Intron | XP_016884952.1 | ||||
XM_017029464.1 | Intron | XP_016884953.1 | ||||
XM_017029465.1 | Intron | XP_016884954.1 | ||||
XM_017029466.1 | Intron | XP_016884955.1 | ||||
XM_017029467.1 | Intron | XP_016884956.1 | ||||
XM_017029468.1 | Intron | XP_016884957.1 | ||||
XM_017029469.1 | Intron | XP_016884958.1 | ||||
XM_017029470.1 | Intron | XP_016884959.1 |