Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGCTCCCTCCTCGTCTCCTTCC[C/G]TTCGTTAATTAGTCCATCGCATTTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603945 | |||||||||||||||||||||||
Literature Links: |
EIF2B5 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
EIF2B5 - eukaryotic translation initiation factor 2B subunit epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003907.2 | 262 | UTR 5 | NP_003898.2 | |||
XM_011513265.1 | 262 | Intron | XP_011511567.1 | |||
XM_011513266.2 | 262 | Intron | XP_011511568.1 |