Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACCGCCTAGACCCAGCCCGCTA[G/C]AGGCAGGCAGTGATGGCTGTGAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf84 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf84 - chromosome 11 open reading frame 84 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138471.2 | 485 | Missense Mutation | CAG,GAG | Q,E 58 | NP_612480.1 | |
XM_005273782.2 | 485 | Missense Mutation | CAG,GAG | Q,E 58 | XP_005273839.1 | |
XM_005273783.2 | 485 | Missense Mutation | CAG,GAG | Q,E 58 | XP_005273840.1 | |
XM_006718437.1 | 485 | Missense Mutation | CAG,GAG | Q,E 58 | XP_006718500.1 | |
XM_011544770.1 | 485 | Missense Mutation | CAG,GAG | Q,E 58 | XP_011543072.1 | |
XM_011544771.1 | 485 | Missense Mutation | CAG,GAG | Q,E 58 | XP_011543073.1 | |
XM_011544772.1 | 485 | Missense Mutation | CAG,GAG | Q,E 58 | XP_011543074.1 |