Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAAAATTGTTTCAAGGCACTTA[A/C]GGCTGCCTAAAAATGAAAACAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605920 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STAU2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STAU2 - staufen double-stranded RNA binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164380.1 | 1272 | Missense Mutation | GTA,TTA | V,L 513 | NP_001157852.1 | |
NM_001164381.1 | 1272 | Missense Mutation | GTA,TTA | V,L 481 | NP_001157853.1 | |
NM_001164382.1 | 1272 | Missense Mutation | GTA,TTA | V,L 447 | NP_001157854.1 | |
NM_001164383.1 | 1272 | Missense Mutation | GTA,TTA | V,L 341 | NP_001157855.1 | |
NM_001164384.1 | 1272 | Intron | NP_001157856.1 | |||
NM_001164385.1 | 1272 | Intron | NP_001157857.1 | |||
NM_014393.2 | 1272 | Intron | NP_055208.2 |
STAU2-AS1 - STAU2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |