Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAACTATAGCCTCACTTCTGAGA[C/G]AGCAGTACTGGTTGCACACACACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600246 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELK4 - ELK4, ETS transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001973.3 | Intron | NP_001964.2 | ||||
NM_021795.2 | Intron | NP_068567.1 | ||||
XM_005244950.4 | Intron | XP_005245007.1 | ||||
XM_005244951.4 | Intron | XP_005245008.1 | ||||
XM_017000580.1 | Intron | XP_016856069.1 |
MFSD4A - major facilitator superfamily domain containing 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181644.4 | Intron | NP_857595.3 | ||||
XM_011509229.2 | Intron | XP_011507531.1 |