Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGATGCCACAGCCCAGAGCCTG[G/A]TGGTGGCTGAGGCTGATAGATTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTNL10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTNL10 - butyrophilin like 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287262.1 | 620 | Missense Mutation | ACC,ATC | T,I 156 | NP_001274191.1 | |
XM_011544015.2 | 620 | Missense Mutation | ACC,ATC | T,I 156 | XP_011542317.1 | |
XM_016999986.1 | 620 | Missense Mutation | ACC,ATC | T,I 156 | XP_016855475.1 | |
XM_016999987.1 | 620 | Missense Mutation | ACC,ATC | T,I 156 | XP_016855476.1 |