Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTACCACCCCAAAGCCTGCAGGCC[A/C]CATACCTGAGTATGCTTACGGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600875 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACYP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACYP1 - acylphosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001107.4 | Intron | NP_001098.1 | ||||
NM_001302616.1 | Intron | NP_001289545.1 | ||||
NM_001302617.1 | Intron | NP_001289546.1 |
ZC2HC1C - zinc finger C2HC-type containing 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |