Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCCTCGGAGTGGGGTCCCAGTC[C/T]GGAGTCTTCAGGTCAGTGGCAGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616658 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf70 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C19orf70 - chromosome 19 open reading frame 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308240.1 | 1028 | UTR 3 | NP_001295169.1 | |||
NM_205767.2 | 1028 | UTR 3 | NP_991330.1 | |||
XM_011527675.2 | 1028 | Intron | XP_011525977.1 | |||
XM_017026248.1 | 1028 | UTR 3 | XP_016881737.1 |
HSD11B1L - hydroxysteroid 11-beta dehydrogenase 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107985320 - uncharacterized LOC107985320 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017027552.1 | 1028 | Intron | XP_016883041.1 | |||
XM_017027553.1 | 1028 | Intron | XP_016883042.1 | |||
XM_017027554.1 | 1028 | Intron | XP_016883043.1 | |||
XM_017027555.1 | 1028 | Intron | XP_016883044.1 |