Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATAGATGGATTCTTGATGTTATT[A/G]TATGTCATTCTTAATGAAAATGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf28 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf28 - chromosome 14 open reading frame 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017923.1 | 464 | Silent Mutation | TTA,TTG | L,L 66 | NP_001017923.1 | |
XM_011536408.1 | 464 | Intron | XP_011534710.1 |
LOC101927418 - uncharacterized LOC101927418 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |