Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGCCTGGGATGGTGCCTCGGCT[A/G]TCTACATCAAGATGAGTCCAGAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604487 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTOG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OTOG - otogelin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277269.1 | 805 | Missense Mutation | ATC,GTC | I,V 269 | NP_001264198.1 | |
NM_001292063.1 | 805 | Missense Mutation | ATC,GTC | I,V 257 | NP_001278992.1 |