Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTGGACTGGAAGACCCCAGAGC[A/G]AGGTAAGACTCACAGACCTAGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300924 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AWAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AWAT1 - acyl-CoA wax alcohol acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013579.2 | 223 | Missense Mutation | CAA,CGA | Q,R 61 | NP_001013597.1 | |
XM_017029295.1 | 223 | Missense Mutation | CAA,CGA | Q,R 61 | XP_016884784.1 |