Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTTCATTTGTGGCTTTAACAGG[C/T]AATTTTAAGGCTTAAAATATTAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TECRL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TECRL - trans-2,3-enoyl-CoA reductase-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010874.4 | Intron | NP_001010874.2 | ||||
XM_005265662.4 | Intron | XP_005265719.1 | ||||
XM_005265663.2 | Intron | XP_005265720.1 | ||||
XM_005265664.2 | Intron | XP_005265721.1 | ||||
XM_005265665.3 | Intron | XP_005265722.1 | ||||
XM_017007959.1 | Intron | XP_016863448.1 | ||||
XM_017007960.1 | Intron | XP_016863449.1 |