Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608011 MIM: 606083 | ||||||||||||||||||||
Literature Links: |
GLT8D1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLT8D1 - glycosyltransferase 8 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GNL3 - G protein nucleolar 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014366.4 | 136 | UTR 5 | NP_055181.3 | |||
NM_206825.1 | 136 | UTR 5 | NP_996561.1 | |||
NM_206826.1 | 136 | UTR 5 | NP_996562.1 |
SNORD136 - small nucleolar RNA, C/D box 136 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD19 - small nucleolar RNA, C/D box 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD19B - small nucleolar RNA, C/D box 19B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD69 - small nucleolar RNA, C/D box 69 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |