Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTACAGATGATATGATATAAAA[C/T]GCAGGGGGTTTTCTTTGGATCAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611342 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf64 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf64 - chromosome 9 open reading frame 64 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317997.1 | 1305 | UTR 3 | NP_001304926.1 | |||
NM_032307.4 | 1305 | UTR 3 | NP_115683.3 | |||
XM_005252271.4 | 1305 | Intron | XP_005252328.1 |