Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAGCAGAGCCCATCCCTCTACC[C/T]GCTCCCTCCTACTCCGGCTTCTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608116 MIM: 615340 | ||||||||||||||||||||
Literature Links: |
HHATL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HHATL - hedgehog acyltransferase-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020707.3 | 1740 | UTR 3 | NP_065758.3 | |||
XM_006713274.2 | 1740 | UTR 3 | XP_006713337.1 | |||
XM_006713275.2 | 1740 | UTR 3 | XP_006713338.1 | |||
XM_011533969.2 | 1740 | UTR 3 | XP_011532271.1 | |||
XM_011533970.2 | 1740 | UTR 3 | XP_011532272.1 | |||
XM_017006935.1 | 1740 | UTR 3 | XP_016862424.1 | |||
XM_017006936.1 | 1740 | UTR 3 | XP_016862425.1 | |||
XM_017006937.1 | 1740 | UTR 3 | XP_016862426.1 | |||
XM_017006938.1 | 1740 | UTR 3 | XP_016862427.1 |
KLHL40 - kelch like family member 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152393.3 | 1740 | Intron | NP_689606.2 |