Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGCCATTCTGGAGAATGAAGAC[C/T]GGATCGAAGATGCCTCGTAAGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615949 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM98 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM98 - transmembrane protein 98 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033504.1 | 629 | Missense Mutation | CGG,TGG | R,W 83 | NP_001028676.1 | |
NM_001301746.1 | 629 | Missense Mutation | CGG,TGG | R,W 83 | NP_001288675.1 | |
NM_015544.2 | 629 | Missense Mutation | CGG,TGG | R,W 83 | NP_056359.2 |