Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGCAGGTGAGCGTCCCCGGTCG[C/T]AGGCCTGCGGATTGGGCAGGGCTTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610885 MIM: 614854 MIM: 611837 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EME1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EME1 - essential meiotic structure-specific endonuclease 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166131.1 | Intron | NP_001159603.1 | ||||
NM_152463.2 | Intron | NP_689676.2 | ||||
XM_005257081.3 | Intron | XP_005257138.1 | ||||
XM_011524395.2 | Intron | XP_011522697.1 | ||||
XM_017024236.1 | Intron | XP_016879725.1 |
LRRC59 - leucine rich repeat containing 59 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL27 - mitochondrial ribosomal protein L27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016504.2 | Intron | NP_057588.1 |
Set Membership: |
HapMap |