Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTTGCTGTTTCTACACTTGCTA[A/G]GGATAGATCCAGCACTGATAACAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616426 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP192 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP192 - centrosomal protein 192 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032142.3 | 275 | Missense Mutation | AAG,AGG | K,R 47 | NP_115518.3 | |
XM_005258107.3 | 275 | Missense Mutation | AAG,AGG | K,R 47 | XP_005258164.1 | |
XM_005258109.3 | 275 | Missense Mutation | AAG,AGG | K,R 47 | XP_005258166.1 | |
XM_005258110.1 | 275 | Intron | XP_005258167.1 | |||
XM_006722326.3 | 275 | Missense Mutation | AAG,AGG | K,R 47 | XP_006722389.1 | |
XM_006722327.3 | 275 | Missense Mutation | AAG,AGG | K,R 47 | XP_006722390.1 | |
XM_006722330.3 | 275 | Missense Mutation | AAG,AGG | K,R 47 | XP_006722393.1 | |
XM_011525673.2 | 275 | Missense Mutation | AAG,AGG | K,R 47 | XP_011523975.1 | |
XM_011525675.2 | 275 | Intron | XP_011523977.1 | |||
XM_017025803.1 | 275 | UTR 5 | XP_016881292.1 | |||
XM_017025804.1 | 275 | Missense Mutation | AAG,AGG | K,R 47 | XP_016881293.1 | |
XM_017025805.1 | 275 | Intron | XP_016881294.1 | |||
XM_017025806.1 | 275 | Intron | XP_016881295.1 | |||
XM_017025807.1 | 275 | Intron | XP_016881296.1 |