Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615943 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGI3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAGI3 - membrane associated guanylate kinase, WW and PDZ domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142782.1 | Intron | NP_001136254.1 | ||||
NM_152900.2 | Intron | NP_690864.2 | ||||
XM_005270737.3 | Intron | XP_005270794.1 | ||||
XM_011541208.2 | Intron | XP_011539510.1 | ||||
XM_017000974.1 | Intron | XP_016856463.1 | ||||
XM_017000975.1 | Intron | XP_016856464.1 |