Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAAAATGAATATATTATTCTAGG[G/T]TAATTTTTTTCCATTCAAATGTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612175 | ||||||||||||||||||||
Literature Links: |
CAB39L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAB39L - calcium binding protein 39 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079670.2 | 1368 | UTR 3 | NP_001073138.1 | |||
NM_001287337.1 | 1368 | UTR 3 | NP_001274266.1 | |||
NM_001287338.1 | 1368 | UTR 3 | NP_001274267.1 | |||
NM_001287339.1 | 1368 | UTR 3 | NP_001274268.1 | |||
NM_030925.3 | 1368 | UTR 3 | NP_112187.2 |