Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTCCTGATGAACAAAGGCTGAAT[C/G]TTAGACTTTCAGAGAAAGATGCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611149 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC30A7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC30A7 - solute carrier family 30 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144884.1 | Intron | NP_001138356.1 | ||||
NM_133496.4 | Intron | NP_598003.2 | ||||
XM_011540779.2 | Intron | XP_011539081.1 | ||||
XM_017000400.1 | Intron | XP_016855889.1 | ||||
XM_017000401.1 | Intron | XP_016855890.1 |