Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTCTGTAGGCAATCAGATGTCA[C/T]TGAAACATTAAACAGGGAAATGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300075 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS6KA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPS6KA3 - ribosomal protein S6 kinase A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004586.2 | Intron | NP_004577.1 | ||||
XM_005274573.2 | Intron | XP_005274630.1 | ||||
XM_005274577.2 | Intron | XP_005274634.1 | ||||
XM_006724507.3 | Intron | XP_006724570.1 | ||||
XM_011545555.1 | Intron | XP_011543857.1 | ||||
XM_011545556.1 | Intron | XP_011543858.1 | ||||
XM_011545557.2 | Intron | XP_011543859.1 | ||||
XM_011545558.2 | Intron | XP_011543860.1 | ||||
XM_011545559.1 | Intron | XP_011543861.1 | ||||
XM_011545560.1 | Intron | XP_011543862.1 | ||||
XM_011545561.1 | Intron | XP_011543863.1 | ||||
XM_011545562.1 | Intron | XP_011543864.1 | ||||
XM_011545563.2 | Intron | XP_011543865.1 | ||||
XM_017029713.1 | Intron | XP_016885202.1 | ||||
XM_017029714.1 | Intron | XP_016885203.1 | ||||
XM_017029715.1 | Intron | XP_016885204.1 | ||||
XM_017029716.1 | Intron | XP_016885205.1 | ||||
XM_017029717.1 | Intron | XP_016885206.1 | ||||
XM_017029718.1 | Intron | XP_016885207.1 | ||||
XM_017029719.1 | Intron | XP_016885208.1 |