Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCCGAGGGCAGCTAGGGGACTA[C/T]GCGGTGGGGCTGGGGGGTCTCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615765 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC16A11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC16A11 - solute carrier family 16 member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153357.1 | 1254 | Missense Mutation | NP_699188.1 | |||
XM_005256488.3 | 1254 | Missense Mutation | XP_005256545.2 | |||
XM_017024281.1 | 1254 | Missense Mutation | XP_016879770.1 | |||
XM_017024282.1 | 1254 | Intron | XP_016879771.1 |
SLC16A13 - solute carrier family 16 member 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |