Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCAATTAAGTTTCTGAACATAC[C/T]TTTGCTGATTCCTACCCTGCAGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXO5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXO5 - exonuclease 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022774.1 | 884 | Missense Mutation | CCT,CTT | P,L 151 | NP_073611.1 | |
XM_005271125.3 | 884 | Missense Mutation | CCT,CTT | P,L 151 | XP_005271182.1 | |
XM_005271126.3 | 884 | Missense Mutation | CCT,CTT | P,L 151 | XP_005271183.1 | |
XM_005271128.3 | 884 | Missense Mutation | CCT,CTT | P,L 151 | XP_005271185.1 | |
XM_011541970.2 | 884 | Missense Mutation | CCT,CTT | P,L 151 | XP_011540272.1 | |
XM_017002099.1 | 884 | Missense Mutation | CCT,CTT | P,L 151 | XP_016857588.1 | |
XM_017002100.1 | 884 | Missense Mutation | CCT,CTT | P,L 151 | XP_016857589.1 | |
XM_017002101.1 | 884 | Missense Mutation | CCT,CTT | P,L 151 | XP_016857590.1 | |
XM_017002102.1 | 884 | Missense Mutation | CCT,CTT | P,L 151 | XP_016857591.1 | |
XM_017002103.1 | 884 | Missense Mutation | CCT,CTT | P,L 151 | XP_016857592.1 | |
XM_017002104.1 | 884 | Missense Mutation | CCT,CTT | P,L 151 | XP_016857593.1 |