Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGTTCCACCGGGCCGTGGAACA[A/G]CACAATGGCAAGACCATTTTCGCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616967 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0753 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KIAA0753 - KIAA0753 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014804.2 | 244 | Intron | NP_055619.2 | |||
XM_006721612.2 | 244 | Intron | XP_006721675.1 | |||
XM_011524090.2 | 244 | Intron | XP_011522392.1 | |||
XM_011524091.2 | 244 | Intron | XP_011522393.1 | |||
XM_011524095.2 | 244 | Intron | XP_011522397.1 | |||
XM_011524096.1 | 244 | Intron | XP_011522398.1 | |||
XM_011524098.2 | 244 | Intron | XP_011522400.1 | |||
XM_011524099.2 | 244 | Intron | XP_011522401.1 | |||
XM_011524100.2 | 244 | Intron | XP_011522402.1 | |||
XM_017025455.1 | 244 | Intron | XP_016880944.1 | |||
XM_017025456.1 | 244 | Intron | XP_016880945.1 | |||
XM_017025457.1 | 244 | Intron | XP_016880946.1 |
MED31 - mediator complex subunit 31 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TXNDC17 - thioredoxin domain containing 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032731.3 | 244 | Silent Mutation | CAA,CAG | Q,Q 21 | NP_116120.1 |
Set Membership: |
HapMap |