Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCATGGTGCACAAATTCATACT[G/T]CTCACTGGTTTGCACCATTCCACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176882 MIM: 602853 | ||||||||||||||||||||
Literature Links: |
PTPRB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTPRB - protein tyrosine phosphatase, receptor type B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTPRR - protein tyrosine phosphatase, receptor type R | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207015.1 | 1916 | Missense Mutation | AAG,CAG | K,Q 524 | NP_001193944.1 | |
NM_001207016.1 | 1916 | Missense Mutation | AAG,CAG | K,Q 430 | NP_001193945.1 | |
NM_002849.3 | 1916 | Missense Mutation | AAG,CAG | K,Q 636 | NP_002840.2 | |
NM_130846.2 | 1916 | Missense Mutation | AAG,CAG | K,Q 391 | NP_570897.2 | |
XM_011538615.2 | 1916 | Missense Mutation | AAG,CAG | K,Q 628 | XP_011536917.1 |