Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCGAGGGCTCCGCATCGCAACCA[A/T]GGCGCTGCGCTGGGGCATCGTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606377 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHDH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHDH - dihydrodiol dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014475.3 | 42 | Missense Mutation | AAG,ATG | K,M 1 | NP_055290.1 | |
XM_005258748.4 | 42 | Intron | XP_005258805.1 | |||
XM_017026598.1 | 42 | Intron | XP_016882087.1 |